Learn More
Molecular Devices Molecular Devices FLIPR Calcium Assay Kit, 2-Plate Sample, Fluorescence, Homogeneous
Catalog No. 5000014483
Encompass_Preferred
Sample kit for performing fluorescence-based intracellular calcium flux assays. The reagents are formulated for a homogeneous mix-and-read workflow to accelerate assay development and screening of GPCR and ion channel targets on compatible instruments.
  • Sample kit containing two Component A vials, each sufficient for one microplate.
  • Fluorescence-based, homogeneous assay format for calcium flux detection.
  • Optimized for GPCR and ion channel screening and pharmacology studies.
  • Compatible with common FLIPR and plate reader systems, including Tetra, Penta, and FlexStation platforms.
  • Component B dilution/assay buffer is sold separately and required for some kit sizes.
Catalog No. 50-000-14483 Supplier Molecular Devices Supplier No. R6133
Sample kit for performing fluorescence-based intracellular calcium flux assays. The reagents are formulated for a homogeneous mix-and-read workflow to accelerate assay development and screening of GPCR and ion channel targets on compatible instruments.
  • Sample kit containing two Component A vials, each sufficient for one microplate.
  • Fluorescence-based, homogeneous assay format for calcium flux detection.
  • Optimized for GPCR and ion channel screening and pharmacology studies.
  • Compatible with common FLIPR and plate reader systems, including Tetra, Penta, and FlexStation platforms.
  • Component B dilution/assay buffer is sold separately and required for some kit sizes.