Learn More
AdipoGen QC-107
SDP
Supplier:  AdipoGen CDXC0136M100
Encompass_Preferred
Chemical. CAS 69352-04-1. Formula C9H10ClIO. MW 296.5. Synthetic. A Hypervalent iodine reagent for oxidation reactions.
Catalog No. 50-256-1460