Learn More
IVF STORE QC-Beads™ For Use as Controls in Counting Sperm Using Manual and Automated Methods
Catalog No. NC3365081
Encompass
Suspensions of latex beads for quality control of counting sperm using manual and automated methods.For in vitro diagnostic use only (FDA Approved)FeaturesTwo different known concentrations of latex beads suspended in an aqueous solutionVolume: 4 ml per vialRoom temperature storageReady to use24-month shelf life
Catalog No. NC3365081 Supplier IVF STORE Supplier No. 0006020
Suspensions of latex beads for quality control of counting sperm using manual and automated methods.For in vitro diagnostic use only (FDA Approved)FeaturesTwo different known concentrations of latex beads suspended in an aqueous solutionVolume: 4 ml per vialRoom temperature storageReady to use24-month shelf life