Learn More
Grainger Micro 100 Boring and Profiling Tool, Solid Carbide, AlTiN, 0.1875 in. Shank, 1 ea
Catalog No. 502754721
Encompass_Preferred
This solid carbide boring and profiling tool is designed for precise axial profiling and boring applications. Engineered for performance in demanding environments, this tool is suitable for use with a variety of materials including alloy steels, stainless steels, tool steels, nickel alloys, and titaniums.
  • Constructed from durable solid carbide for enhanced tool life and performance.
  • Features an AlTiN coating for improved wear resistance and heat management.
  • Designed for right-hand cutting applications.
  • Includes a 0.005 in. corner radius for detailed profiling tasks.
  • Shank diameter of 0.1875 in. with a maximum bore depth of 0.400 in.
Catalog No. 50-275-4721 Supplier Grainger Supplier No. 15T987
This solid carbide boring and profiling tool is designed for precise axial profiling and boring applications. Engineered for performance in demanding environments, this tool is suitable for use with a variety of materials including alloy steels, stainless steels, tool steels, nickel alloys, and titaniums.
  • Constructed from durable solid carbide for enhanced tool life and performance.
  • Features an AlTiN coating for improved wear resistance and heat management.
  • Designed for right-hand cutting applications.
  • Includes a 0.005 in. corner radius for detailed profiling tasks.
  • Shank diameter of 0.1875 in. with a maximum bore depth of 0.400 in.