Learn More
Bioworld QC Buffer, 250 mL
Catalog No. 50196888
Encompass_Preferred
Product DescriptionQC buffer is a plasmid DNA purification buffer. It is composed of 1.0M NaCl, 50 mM MOPS, 15% isopropanol and has a pH value of 7.0.
Catalog No. 50-196-888 Supplier Bioworld Supplier No. 107600361
Product DescriptionQC buffer is a plasmid DNA purification buffer. It is composed of 1.0M NaCl, 50 mM MOPS, 15% isopropanol and has a pH value of 7.0.