Learn More
Tecan Range C Dye for QC Kit
Catalog No. NC1321975
Encompass_Preferred
96-well volume range: 2. 0-9. 99 L, 384-well volume range: 0. 5-2. 499 L. 1 bottle/sales unit (110 mL). The Tecan QC Kit is a quality control tool for pipetting volume verification of your liquid handling platform, ensuring full regulatory compliance.
This item is not returnable. View return policy
Catalog No. NC1321975 Supplier Tecan Supplier No. 30062842
96-well volume range: 2. 0-9. 99 L, 384-well volume range: 0. 5-2. 499 L. 1 bottle/sales unit (110 mL). The Tecan QC Kit is a quality control tool for pipetting volume verification of your liquid handling platform, ensuring full regulatory compliance.