Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ HemoScreen QC Kit

Catalog No. 23123807
Change view
Click to view available options
Quantity:
3 Levels/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-123-807 3 Levels/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-123-807 Supplier R&D Systems™ Supplier No. HSFA0020001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Customized HemoScreen Quality Control Kit used to evaluate system accuracy and precision.

  • Each control kit contains 3 levels, 2 vials each.
  • HemoScreen also includes extensive internal QC.

Specifications

Certifications/Compliance FDA Licensed Tests
Detectable Analytes Blood, Red Blood Cells, Hemoglobin, Platelets, White Blood Cells
For Use With (Equipment) PixCell™ HemoScreen
Sample Type Whole Blood, Serum, Plasma
Type QC Kit
Level 3
Quantity 3 Levels/Box
Source Human Erythrocytes, Simulated Leukocytes and Platelets in Plasma-like Fluid with Preservatives
Stability Stable Through Expiration Date
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.