Learn More
ERA WATERS CORP QC METALS
SDP
Catalog No. NC9754230
Encompass
One 15 mL screw-cap vial yields up to 2 liters after dilution. Use with ICP-OES, ICP-MS and AA methods.
Catalog No. NC9754230 Supplier ERA WATERS CORP Supplier No. 697
One 15 mL screw-cap vial yields up to 2 liters after dilution. Use with ICP-OES, ICP-MS and AA methods.