Learn More
Tosoh Bioscience LLC QC-PAK GFC 200 7.8X150
Supplier:  Tosoh Bioscience LLC 0016215
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel QC-PAK GFC 200; Media: Silica; Particle: 5 micrometer; Pore Size: 125 angstrom; 7.8mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-405