Learn More
SCP Science QC STANDARD NO4
Catalog No. NC9212571
Encompass
Quality Control Standard 4, Designed Specifically for EPA 200.7, 200.8, 6010 and Superfund CLP, Traceability to NIST
Catalog No. NC9212571 Supplier SCP Science Supplier No. 140102041
Quality Control Standard 4, Designed Specifically for EPA 200.7, 200.8, 6010 and Superfund CLP, Traceability to NIST