Learn More
QC-Standard Solution, For EN ISO 9377-2, Analytical Standard, MilliporeSigma™ Supelco™
Catalog No. 111016512 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
11-101-6512 1 mL
1 options
Catalog No. 11-101-6512 Supplier MilliporeSigma Supelco Supplier No. 517061MLF
Specifications
Description Contains Mineral oil type A and B, ˜5 mg/mL each, BAM reference material 5002 and 5003 in n-Heptane
Quantity 1 mL
Grade Analytical Standard