Learn More
ERA WATERS CORP QC WASTE WATER HARDNESS
SDP
Catalog No. NC9624258
Encompass
Waste Water Hardness ,One 500 mL bottle to be analyzed for calcium, magnesium, total hardness as CaCO3,calcium hardness as CaCO3 and total suspended solids
Catalog No. NC9624258 Supplier ERA WATERS CORP Supplier No. 507
Waste Water Hardness ,One 500 mL bottle to be analyzed for calcium, magnesium, total hardness as CaCO3,calcium hardness as CaCO3 and total suspended solids