Learn More
ERA WATERS CORP QC WASTEWATER COMPLEX NUTRIENT
SDP
Catalog No. NC9409742
Encompass
One 15 mL screw-cap vial yields up to 2 liters after dilution.
Catalog No. NC9409742 Supplier ERA WATERS CORP Supplier No. 525
One 15 mL screw-cap vial yields up to 2 liters after dilution.