Learn More
TARGETMOL CHEMICALS INC QC1 5MG
SDP
Catalog No. 502580889
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Qc1 is an inhibitor of threonine dehydrogenase (TDH) inhibitor and can be used in studies about metabolic diseases. purity: 100%
Catalog No. 50-258-0889 Supplier TARGETMOL CHEMICALS INC Supplier No. T232095MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Qc1 is an inhibitor of threonine dehydrogenase (TDH) inhibitor and can be used in studies about metabolic diseases. purity: 100%