Learn More
Medchemexpress LLC HY-104048 1mg Medchemexpress, QC6352 CAS:1851373-36-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1040481MG
Encompass_Preferred
Medchemexpress, HY-104048 1mg QC6352 CAS:1851373-36-8 QC6352 is an orally available, selective and potent KDM4C inhibitor with an IC50 of 35 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8742