Learn More
Medchemexpress LLC HY-112609 5mg Medchemexpress, QCA570 CAS:2207569-08-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1126095MG
Encompass_Preferred
Medchemexpress, HY-112609 5mg QCA570 CAS:2207569-08-0 QCA570 is a potent BET degrader based on PROTAC, with an IC50 of 10 nM for BRD4 BD1 Protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0824