Learn More
Agilent Technologies QCKCHANGE LIGHTING ST-DIR MUTA
Catalog No. NC9620881
Encompass
Contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.
Catalog No. NC9620881 Supplier Agilent Technologies Supplier No. 210518
Contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.