Learn More
SPEX CERTIPREP LLC 3P 27 Component ICP Standard in 4% HNO3 + Tr HF, 100 mL
Catalog No. NC1806317
Encompass_Preferred
(HPS by Zeptometrix) ICP Multi-Component Quality Control Standard Set with 27 Components at 100 ug/mL in 4% HNO3+Tr HF.
Catalog No. NC1806317 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCS27100
(HPS by Zeptometrix) ICP Multi-Component Quality Control Standard Set with 27 Components at 100 ug/mL in 4% HNO3+Tr HF.