Learn More
Waters Corp Waters QDa QC Reference Material, 9-component mix, 1 mL, Liquid
Catalog No. 502037960 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The QDa QC Reference Material is a quality control reference material designed to benchmark and assess the performance of a UPLC system, specifically including a mass detector. This 9-component mix provides a comprehensive standard to evaluate chromatographic systems and ensure readiness for sample analysis.
  • 1 mL, 9-component mix for UPLC system performance assessment.
  • Supports ESI (+ and –) and APCI+ detection modes across a wide m/z range.
  • Optimized for equal component response in ESI+ mode.
  • Facilitates system benchmarking, monitoring, and problem identification.
  • Features Reversed Phase separation and MS detection.
  • Liquid formulation; store at 4°C (Fridge) with a 12-month shelf life.
  • Ships ambient; one unit per package.
Catalog No. 50-203-7960 Supplier Waters Corp Supplier No. 186007345
The QDa QC Reference Material is a quality control reference material designed to benchmark and assess the performance of a UPLC system, specifically including a mass detector. This 9-component mix provides a comprehensive standard to evaluate chromatographic systems and ensure readiness for sample analysis.
  • 1 mL, 9-component mix for UPLC system performance assessment.
  • Supports ESI (+ and –) and APCI+ detection modes across a wide m/z range.
  • Optimized for equal component response in ESI+ mode.
  • Facilitates system benchmarking, monitoring, and problem identification.
  • Features Reversed Phase separation and MS detection.
  • Liquid formulation; store at 4°C (Fridge) with a 12-month shelf life.
  • Ships ambient; one unit per package.