Learn More
Waters Corp Waters Aperture Disk Assembly, Performance Mode, 0.2 mm, for QDa
Catalog No. NC0793378 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Aperture Disk Assembly serves as a replacement component for QDa and RADIAN ASAP instruments, specifically when they are configured for Performance Mode operation.
  • Designed as a replacement part.
  • Compatible with QDa and RADIAN ASAP mass spectrometry instruments.
  • Configured for Performance Mode operation.
  • Aperture type.
Catalog No. NC0793378 Supplier Waters Corp Supplier No. 700009768
This Aperture Disk Assembly serves as a replacement component for QDa and RADIAN ASAP instruments, specifically when they are configured for Performance Mode operation.
  • Designed as a replacement part.
  • Compatible with QDa and RADIAN ASAP mass spectrometry instruments.
  • Configured for Performance Mode operation.
  • Aperture type.