Learn More
Izon Science Usa Ltd qEVoriginal/35nm Gen 2 Column, 35 nm pore size, 5-pack, 500 μL sample loading volume, proprietary agarose resin, enhanced separation performance
SDP
Catalog No. NC2396265
Encompass
qEVoriginal/35nm Gen 2 Column, 35 nm pore size, 5-pack, 500 μL sample loading volume, proprietary agarose resin, enhanced separation performance
Catalog No. NC2396265 Supplier Izon Science Usa Ltd Supplier No. ICO35
qEVoriginal/35nm Gen 2 Column, 35 nm pore size, 5-pack, 500 μL sample loading volume, proprietary agarose resin, enhanced separation performance