Learn More
Bioworld QF Buffer, 250 mL
Catalog No. 50196890
Encompass_Preferred
Product Description QF buffer is a plasmid DNA purification buffer. It is composed of 1.25 M NaCl, 50 mM Tris-Cl, 15% isopropanol and has a pH value of 8.5.
Catalog No. 50-196-890 Supplier Bioworld Supplier No. 107600371
Product Description QF buffer is a plasmid DNA purification buffer. It is composed of 1.25 M NaCl, 50 mM Tris-Cl, 15% isopropanol and has a pH value of 8.5.