Learn More
Accuflow Systems Quattroflow Pump Chamber, Polypropylene, 15 mL, Gamma Irradiated
Catalog No. NC2854683
Encompass_Preferred
The Quattroflow QF15DISPP-3-GAMMA single-use pump chamber is a disposable, product-wetted assembly designed for sterile fluid transfer with Quattroflow QF150 single-use pumps. It provides a gamma-irradiated wetted path to minimize cross-contamination risk and remove the need for cleaning and validation between batches.
  • Disposable polypropylene pump housing and valve plate
  • Santoprene diaphragms for flexible sealing
  • EPDM valves and O-rings
  • Gamma irradiatable, maximum dose 50 kGy
  • Filling volume 15 mL without connectors
  • Residual volume approximately 1-3 mL after idle
  • Product-wetted surface area approximately 112 cm²
  • Reduces cleaning, validation, and cross-contamination risk
Catalog No. NC2854683 Supplier Accuflow Systems Supplier No. QF15DISPP3GAMMA
The Quattroflow QF15DISPP-3-GAMMA single-use pump chamber is a disposable, product-wetted assembly designed for sterile fluid transfer with Quattroflow QF150 single-use pumps. It provides a gamma-irradiated wetted path to minimize cross-contamination risk and remove the need for cleaning and validation between batches.
  • Disposable polypropylene pump housing and valve plate
  • Santoprene diaphragms for flexible sealing
  • EPDM valves and O-rings
  • Gamma irradiatable, maximum dose 50 kGy
  • Filling volume 15 mL without connectors
  • Residual volume approximately 1-3 mL after idle
  • Product-wetted surface area approximately 112 cm²
  • Reduces cleaning, validation, and cross-contamination risk