Learn More
MilliporeSigma Q-Gard A Cartridge
Catalog No. NC1786251
Encompass
The Q-Gard A Cartridge is designed to remove ionic contaminants below trace level to match the water quality required for your applications.
Catalog No. NC1786251 Supplier MilliporeSigma Supplier No. CP2ALLREST
The Q-Gard A Cartridge is designed to remove ionic contaminants below trace level to match the water quality required for your applications.