Learn More
INDICAL INC INDISPIN PATHOGEN KIT 50
SDP
Supplier:  INDICAL INC SP54104
Encompass_Preferred
The IndiSpin Pathogen Kit is designed for high-quality and efficient extraction of pathogen nucleic acids, including viral RNA and DNA, and bacterial DNA. It is suitable for use with a wide variety of sample types such as whole blood, serum, plasma, other body fluids, swabs, washes, and tissue homogenates. Key features of the kit include:¿A unified protocol for both DNA and RNA extraction.¿Effective removal of inhibitors and contaminants for high-purity results.¿Compatibility with automation on the QIAcube for low to medium throughput workflows.¿A centrifugation-based protocol for straightforward implementation.The purified nucleic acids are free of proteins, nucleases, and other impurities, making them ideal for downstream applications, including real-time PCR-based pathogen detection. This versatile kit is particularly well-suited for applications in fields such as veterinary diagnostics, where sample types can vary significantly.
Catalog No. NC1684410