Learn More
QIAGEN QIAcard FTA Mini
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
- QIAcard FTA Mini format with 2 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
- FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Specifications
| Description | Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Mini Cards. |
| Isolation Technology | Impregnated Specialized Filter Paper Technology |
| Scale | Mini |
| Sample Type | Blood |
| For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology |
| No. of Samples | 2 |
| Product Type | FTA Mini Card |
| Quantity | 100 Pk. |
| Sample Volume | 125 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.