Learn More
Maine Molecular Quality Controls Inc QIAstat-Dx RP SARS-CoV-2 Control Panel
SDP
Catalog No. NC1990581
Encompass
QIAstat-Dx RP SARS-CoV-2 Control Panel is intended for use as an external positive and negative quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of bacterial and viral pathogens on the QIAstat-Dx Respiratory SARS-CoV-2 Panel performed on QIAstat-Dx systems. QIAstat-Dx RP SARS-CoV-2 Control Panel is composed of synthetic RNA specifically designed for and intended to be used solely with the QIAstat-Dx Respiratory SARS-CoV-2 Panel. QIAstat-Dx RP SARS-CoV-2 Control Panel is composed of 3 controls, QIAstat-Dx RP Positive A, QIAstat-Dx RP Positive B1, and QIAstat-Dx RP Negative. The QIAstat-Dx RP SARS-CoV-2 Control Panel is comprised of 12 tubes, 4 tubes of QIAstat-Dx RP Positive A, 4 tubes of QIAstat-Dx RP Positive B1, and 4 tubes of QIAstat-Dx RP Negative, 300µL each.
Catalog No. NC1990581 Supplier Maine Molecular Quality Controls Inc Supplier No. M360V1.2
QIAstat-Dx RP SARS-CoV-2 Control Panel is intended for use as an external positive and negative quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of bacterial and viral pathogens on the QIAstat-Dx Respiratory SARS-CoV-2 Panel performed on QIAstat-Dx systems. QIAstat-Dx RP SARS-CoV-2 Control Panel is composed of synthetic RNA specifically designed for and intended to be used solely with the QIAstat-Dx Respiratory SARS-CoV-2 Panel. QIAstat-Dx RP SARS-CoV-2 Control Panel is composed of 3 controls, QIAstat-Dx RP Positive A, QIAstat-Dx RP Positive B1, and QIAstat-Dx RP Negative. The QIAstat-Dx RP SARS-CoV-2 Control Panel is comprised of 12 tubes, 4 tubes of QIAstat-Dx RP Positive A, 4 tubes of QIAstat-Dx RP Positive B1, and 4 tubes of QIAstat-Dx RP Negative, 300µL each.