Learn More
MSC Qingdao Preco Imp. & Exp. Co., Ltd. Vernier Caliper, Carbon Steel, 0-80 in, 8 in Jaw, 0.001 in Graduation
Catalog No. 5000511238
Encompass
This heavy-duty vernier caliper is designed for precise measurements in industrial applications. Constructed from carbon steel, it offers a measurement range of 0 to 80 inches with fine adjustment, making it suitable for a variety of tasks requiring accurate diameter and length measurements.
  • Measures outside and inside dimensions
  • Features fine adjustment for precise readings
  • Constructed from carbon steel for durability
  • Offers a measurement range of 0 to 80 inches
  • Provides 0.001 inch / 0.02 mm graduation for accuracy
  • Equipped with 8 inch long jaws
Catalog No. 50-005-11238 Supplier MSC Supplier No. 06411342
This heavy-duty vernier caliper is designed for precise measurements in industrial applications. Constructed from carbon steel, it offers a measurement range of 0 to 80 inches with fine adjustment, making it suitable for a variety of tasks requiring accurate diameter and length measurements.
  • Measures outside and inside dimensions
  • Features fine adjustment for precise readings
  • Constructed from carbon steel for durability
  • Offers a measurement range of 0 to 80 inches
  • Provides 0.001 inch / 0.02 mm graduation for accuracy
  • Equipped with 8 inch long jaws