Learn More
Chemglass Life Sciences Chemglass Quick Connect Set, 3/4 in (M30) 316 Stainless Steel, Female-Female
SDP
Catalog No. NC2895392
Encompass_Preferred
Chemglass CG-1969-M-86 is a 3/4 in (0.75 in) quick connect set fabricated from 316 stainless steel for corrosion resistance and durability. It is used for rapid connection and disconnection of hoses and reactor systems and is suitable for a wide temperature range.
  • 316 stainless steel construction.
  • Fluorosilicone seals for chemical compatibility.
  • Both ends terminate in M30 female fittings.
  • Both male and female couplers have shutoffs.
  • Temperature range: -60 to 200° C.
  • Designed for quick connect/disconnect of hoses and reactor systems.
Catalog No. NC2895392 Supplier Chemglass Life Sciences Supplier No. CG1969M86
Chemglass CG-1969-M-86 is a 3/4 in (0.75 in) quick connect set fabricated from 316 stainless steel for corrosion resistance and durability. It is used for rapid connection and disconnection of hoses and reactor systems and is suitable for a wide temperature range.
  • 316 stainless steel construction.
  • Fluorosilicone seals for chemical compatibility.
  • Both ends terminate in M30 female fittings.
  • Both male and female couplers have shutoffs.
  • Temperature range: -60 to 200° C.
  • Designed for quick connect/disconnect of hoses and reactor systems.