Learn More
Aviva Systems Biology Corp QKI Antibody - N-terminal region (ARP63894_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63894_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against QKI. It was validated on Western Blot. . Alternate names: DKFZp586I0923, Hqk, QK, QK1, QK3, hqkIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-109