Learn More
CPC Scientific H-Gln-Lys-Arg-Pro-Ser-Gln-Arg-Ser-Lys-Tyr-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP041B
Encompass
Bovine myelin basic protein fragment 4-14 described as the most specific substrate for the selective assay of protein kinase C. Its Km value of 7 µM is comparable with the value for H1 histone and intact myelin basic protein, but its velocity of phosphorylation is twice as fast. ONE-LETTER SEQUENCE: QKRPSQRSKYLMOLECULAR FORMULA: C60H103N21O17MOLECULAR WEIGHT:1390.59STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [126768-94-3], RESEARCH AREA: Multiple SclerosisREFERENCES: I. Yasuda et al., BBRC, 166, 1220 (1990)
Catalog No. 50-211-0193