Learn More
Medchemexpress LLC QL-X-138 | 1469988-63-3 | 99.8% | 421.45 | C25H19N5O2 | 1MG
SDP
Catalog No. 5000237046
Encompass_Preferred
QL-X-138 is a small-molecule BTK/MNK dual kinase inhibitor that covalently binds to BTK and non-covalently to MNK. It is provided as a high-purity solid for laboratory research and has been characterized in preclinical studies for activity against hematologic malignancies and dengue virus.
  • Dual inhibition of Bruton tyrosine kinase (BTK) and MAPK-interacting kinase (MNK).
  • Covalent binding to BTK and non-covalent binding to MNK.
  • High-purity solid form suitable for laboratory research.
  • Molecular formula C25H19N5O2 and molecular weight 421.45.
  • Demonstrated preclinical activity in models of lymphoma, leukemia, and dengue virus.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-37046 Supplier Medchemexpress LLC Supplier No. HY1246451MG
QL-X-138 is a small-molecule BTK/MNK dual kinase inhibitor that covalently binds to BTK and non-covalently to MNK. It is provided as a high-purity solid for laboratory research and has been characterized in preclinical studies for activity against hematologic malignancies and dengue virus.
  • Dual inhibition of Bruton tyrosine kinase (BTK) and MAPK-interacting kinase (MNK).
  • Covalent binding to BTK and non-covalent binding to MNK.
  • High-purity solid form suitable for laboratory research.
  • Molecular formula C25H19N5O2 and molecular weight 421.45.
  • Demonstrated preclinical activity in models of lymphoma, leukemia, and dengue virus.
  • For research use only; not for human or clinical use.