Learn More
Medchemexpress LLC QL-X-138 | 1469988-63-3 | MFCD34567267 | 99.8% | 421.45 | C25H19N5O2 | 10MG
SDP
Catalog No. 5000238810
Encompass_Preferred
QL-X-138 is a small-molecule research compound that acts as a potent and selective dual inhibitor of Bruton's tyrosine kinase (BTK) and mitogen-activated protein kinase-interacting kinases (MNK). It binds covalently to BTK and non-covalently to MNK, has reported antiviral activity against dengue virus 2, and is supplied for laboratory research use only.
  • Potent dual inhibition of BTK and MNK.
  • Covalent binding to BTK and non-covalent binding to MNK.
  • Reported antiviral activity against dengue virus 2.
  • High purity (99.77%).
  • Available in small research quantities (1 mg, 5 mg, 10 mg; larger sizes by quote).
  • Recommended storage conditions provided for powder and solutions.
Catalog No. 50-002-38810 Supplier Medchemexpress LLC Supplier No. HY12464510MG
QL-X-138 is a small-molecule research compound that acts as a potent and selective dual inhibitor of Bruton's tyrosine kinase (BTK) and mitogen-activated protein kinase-interacting kinases (MNK). It binds covalently to BTK and non-covalently to MNK, has reported antiviral activity against dengue virus 2, and is supplied for laboratory research use only.
  • Potent dual inhibition of BTK and MNK.
  • Covalent binding to BTK and non-covalent binding to MNK.
  • Reported antiviral activity against dengue virus 2.
  • High purity (99.77%).
  • Available in small research quantities (1 mg, 5 mg, 10 mg; larger sizes by quote).
  • Recommended storage conditions provided for powder and solutions.