Learn More
Medchemexpress LLC QL47B TFA | 96.0% | 992.07 | 1 MG
SDP
Catalog No. 5000394374
Encompass_Preferred
QL47B TFA is a biotinylated analogue of QL47, functioning as a potent inhibitor of BTK with an IC50 value of 1.3 μM. It has demonstrated anti-tumor activity.
  • Potent inhibitor of BTK (IC50 value of 1.3 μM)
  • Biotinylated analogue
  • Exhibits anti-tumor activity
  • Available in various pack sizes
  • Purity of 96.03%
Catalog No. 50-003-94374 Supplier Medchemexpress LLC Supplier No. HY160246A1MG
QL47B TFA is a biotinylated analogue of QL47, functioning as a potent inhibitor of BTK with an IC50 value of 1.3 μM. It has demonstrated anti-tumor activity.
  • Potent inhibitor of BTK (IC50 value of 1.3 μM)
  • Biotinylated analogue
  • Exhibits anti-tumor activity
  • Available in various pack sizes
  • Purity of 96.03%