Learn More
Medchemexpress LLC QL47B TFA | 96.0% | 992.07 | 5 MG
SDP
Catalog No. 5000393973
Encompass_Preferred
QL47B TFA is a biotinylated analogue of QL47 (HY-80003), functioning as a potent inhibitor of BTK with an IC50 value of 1.3 μM. It also exhibits anti-tumor activity.
  • Potent inhibitor of BTK (IC50 value of 1.3 μM).
  • Exhibits anti-tumor activity.
Catalog No. 50-003-93973 Supplier Medchemexpress LLC Supplier No. HY160246A5MG
QL47B TFA is a biotinylated analogue of QL47 (HY-80003), functioning as a potent inhibitor of BTK with an IC50 value of 1.3 μM. It also exhibits anti-tumor activity.
  • Potent inhibitor of BTK (IC50 value of 1.3 μM).
  • Exhibits anti-tumor activity.