Learn More
Medchemexpress LLC Ql9 tfa | 95.91% | 5 MG
SDP
Catalog No. 5000425595
Encompass_Preferred
QL9 (QLSPFPFDL) TFA is a high-affinity alloantigen for the 2C T cell receptor (TCR), intended for research use only.
  • High-affinity alloantigen
  • Specific for the 2C T cell receptor (TCR)
  • Suitable for research applications
Catalog No. 50-004-25595 Supplier Medchemexpress LLC Supplier No. HYP0287A5MG
QL9 (QLSPFPFDL) TFA is a high-affinity alloantigen for the 2C T cell receptor (TCR), intended for research use only.
  • High-affinity alloantigen
  • Specific for the 2C T cell receptor (TCR)
  • Suitable for research applications