Learn More
Cincinati Surgical™ Qlicksmart BladeFlask EVO Single-handed Blade Remover
SDP
Catalog No. 19200289
Change view
Click to view available options
Size:
# 10 to # 27, # 60, # 70
Catalog No. Size
19-200-289 # 10 to # 27, # 60, # 70
1 options
This item is not returnable. View return policy
Catalog No. 19-200-289 Supplier Cincinati Surgical™ Supplier No. 48EVO
Add to Cart
Add to Cart
This item is not returnable. View return policy
Qlicksmart BladeFlask EVO Single-handed Blade Remover removes all standard-sized surgical blades, including size 60 and 70 blades.
Qlicksmart BladeFlask EVO Single-handed Blade Remover removes all standard-sized surgical blades, including size 60 and 70 blades.
  • Can be mounted on wall or countertop
  • Holds 100 blades
Specifications
For Use With (Application) Blade Removal
Size # 10 to # 27, # 60, # 70
For Use With (Equipment) Surgical Blade
Disposable Single-use
Sterility Non-sterile