Learn More
Cincinati Surgical™ Qlicksmart BladeFlask Single-handed Blade Remover
SDP
Catalog No. 19200290
Change view
Click to view available options
Size:
# 6 to # 26
Catalog No. Size
19-200-290 # 6 to # 26
1 options
This item is not returnable. View return policy
Catalog No. 19-200-290 Supplier Cincinati Surgical™ Supplier No. 48QS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Qlicksmart BladeFlask Single-handed Blade Remover removes standard-sized surgical blades.
Qlicksmart BladeFlask Single-handed Blade Remover removes standard-sized surgical blades sizes 6 through 26.
  • Can be mounted on wall or countertop
  • Holds 100 blades
Specifications
For Use With (Application) Blade Removal
Size # 6 to # 26
For Use With (Equipment) Surgical Blade
Disposable Single-use
Sterility Non-sterile