Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ QMS™ Everolimus Immunoassays, Calibrators and Controls

Catalog No. 23278468 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Everolimus Calibrator Set
Everolimus Control Set
Everolimus Immunoassay
Description:
For sale in the U.S.A., Mexico and Brazil.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Description
23-278-468 Everolimus Control Set -
23-278-443 Everolimus Immunoassay For sale in the U.S.A., Mexico and Brazil.
23-278-440 Everolimus Calibrator Set For sale in the U.S.A., Mexico and Brazil.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-278-468 Supplier Thermo Scientific™ Supplier No. 0373878
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain results you can count on for immunosuppressant drug monitoring with the Thermo Scientific™ QMS™ Everolimus Immunoassay.

Obtain results you can always count on for the crucial monitoring of everolimus drug levels in kidney and liver transplant patients with the Thermo Scientific™ QMS™ Everolimus Immunoassay. Using our proprietary QMS technology, these liquid, ready-to-use reagents provide excellent accuracy and precision. Applications are available on our compact Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.

Key Features:
  • Optimal low-end sensitivity with a broad dynamic range
  • Excellent correlation with HPLC-MS, the gold standard method
  • Precise throughout the standard assay range
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Everolimus
Sample Type Whole Blood
Type Everolimus Control Set
Quantity 3 levels, 1 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.