Description
Achieve high sensitivity and excellent low-end precision when monitoring kidney, liver and heart transplant patients with the Thermo Scientific™ QMS™ Tacrolimus Immunoassay. With a reportable range down to 1ng/mL, our Tacrolimus Immunoassay is the perfect aid for monitoring patients on multiple drug therapies.The reagents are liquid and ready-to-use with applications available on a wide variety of other laboratory instrumentation. List of applications available upon request.
- ISD Assays utilize a recombinant DNA technology to produce a unique homogeneous enzyme immunoassay system
- QMS ISD and TDM Assays use coated microparticle beads
- Technology is based on competitive inhibition principles and photometrically measures rate of absorbance change
Accurate:
- Excellent low-end sensitivity
- Correlation with gold standard methods
- Precise throughput standard curve assay range
- Lot-to-lot consistency
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Detectable Analytes | Tacrolimus |
| Sample Type | Whole Blood |
| Type | Tacrolimus Calibrators |
| Quantity | 6 calibrator set: 4 mL Cal A, 2 mL Cal B to E |