Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Controls

Catalog No. 23279111 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Lamotrigine Controls
Topiramate Controls
Zonisamide Controls
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
23-279-111 Zonisamide Controls
23-279-105 Lamotrigine Controls
23-279-109 Topiramate Controls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-279-111 Supplier Thermo Scientific™ Supplier No. 0373373
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.

Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Zonisamide Controls
Concentration 8.0, 25.0, 50.0 μg/mL
Detectable Analytes Zonisamide
Sample Type Plasma, Serum
Type Control
Level 3
Quantity 3 x 2.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.