Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators

Catalog No. p-6158072
Change view
Click to view available options
Detectable Analytes:
Gentamicin
Tobramycin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-278-447 Tobramycin
23-278-446 Gentamicin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-447 Supplier Thermo Scientific™ Supplier No. 0374116
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ TDM Calibrators, offered in Levels A to F.

Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.

Reduces Labor
  • Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
  • Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
Ready-to-use
  • Liquid stable and ready-to-use
  • Applicable to general chemistry analyzers
  • Validated parameters assure lab-to-lab reproducibility
  • Superb reagent stability
Optimal Sensitivity
  • Excellent correlation to reference methods
  • Low cross-reactivity with metabolites and commonly co-administered drugs
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Tobramycin
Sample Type Plasma, Serum
Type Calibrator
Quantity Levels A-F, 1 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.