Learn More
TARGETMOL CHEMICALS INC QN523 5MG
SDP
Catalog No. 502579363
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. QN523 represents a novel scaffold with drug-like properties showing potent in vitro cytotoxicity in a panel of 12 cancer cell lines. QN523 shows significant in vivo efficacy in a pancreatic cancer xenograft model. Autophagy is a major mechanism of action. purity: 97%
Catalog No. 50-257-9363 Supplier TARGETMOL CHEMICALS INC Supplier No. T643745MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. QN523 represents a novel scaffold with drug-like properties showing potent in vitro cytotoxicity in a panel of 12 cancer cell lines. QN523 shows significant in vivo efficacy in a pancreatic cancer xenograft model. Autophagy is a major mechanism of action. purity: 97%