Learn More
TARGETMOL CHEMICALS INC QNZ 10MG
SDP
Catalog No. 502579489
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. QNZ (EVP4593) (EVP4593) is an effective inhibitor of NF-(kappa)B activation and TNF-(alpha) production. The IC50 of QNZ for NF-(kappa)B and TNF-(alpha) is11 nM and 7 nM respectively. purity: 99%
Catalog No. 50-257-9489 Supplier TARGETMOL CHEMICALS INC Supplier No. T620410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. QNZ (EVP4593) (EVP4593) is an effective inhibitor of NF-(kappa)B activation and TNF-(alpha) production. The IC50 of QNZ for NF-(kappa)B and TNF-(alpha) is11 nM and 7 nM respectively. purity: 99%