Learn More
Enzo Life Sciences QNZ (1mg). CAS: 545380-34-5
SDP
Supplier:  Enzo Life Sciences BMLEI3520001
Encompass_Preferred
Inhibitor of NF-κB which displays potent inhibitory effects on both NF-κB transcriptional activation (IC50=11nM) and TNF-α production (IC50= 7nM). A useful tool to explore the role of NF-κB in cellular signaling pathways. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (20 mg/ml); DMF (30mg/ml); 100% ethanol (10 mg/ml); not soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0738