Learn More
Abcam QNZ, NF-kB inhibitor, 1MG
Catalog No. 502706505
Encompass_Preferred
MW 358.4 Da, Purity >98%. Potent, selective NF-kB inhibitor (IC₅₀ = 11 nM). Inhibits LPS-induced TNFα production (IC₅₀ = 7 nM). Reduces edema in inflammation in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6505 Supplier Abcam Supplier No. AB1415881MG
MW 358.4 Da, Purity >98%. Potent, selective NF-kB inhibitor (IC₅₀ = 11 nM). Inhibits LPS-induced TNFα production (IC₅₀ = 7 nM). Reduces edema in inflammation in vivo..
The product is subject to the following: Abcam Restricted Use Statement