Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black PVC Lined Cap for 2.5-Gallon HDPE square Hedpak

Catalog No. 02991093
Change view
Click to view available options
Thread Size (Metric):
AP mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
02-991-093 AP mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-991-093 Supplier Qorpak™ Supplier No. CAP05976

missing translation for 'validMainframeMsg'

Accessory for HDPE Square Hedpak

  • Available in ribbed style
  • Thread finish: 51mm-400

Specifications

Color Black
Material Polypropylene
No. per Case 800 Cs.
Thread Size (Metric) AP mm
Liner Material PVC
Neck Finish 51-400
For Use With (Equipment) Carboy
Product Type Ribbed Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.