Learn More
Qorpak™ Clear Graduated Medium Round Ready to Clean Bottles
Description
- Made of glass
- Wide mouth
- Graduated in both milliliters and ounces
- Multipurpose containers for use with biological and pathological specimens and for storing industrial laboratory chemicals and reagents
- With black polypropylene caps and PTFE discs
- Available in volumes ranging from 1 to 8 oz. (30 to 240mL)
Specifications
Specifications
| Capacity | 2 oz., 60 mL |
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Clear |
| Diameter (English) | 1.65 in. |
| Material | Glass |
| Packaging | Conventional Pack |
| Quantity | 48/Cs. |
| Sterility | Non-sterile |
| Closure Color | Black |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.