Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Straight Sided Round Bottles with Green Thermoset F217 and PTFE Caps

Catalog No. 02992428
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
120 mL
180 mL
240 mL
480 mL
960 mL
Quantity:
12/Cs.
144/Cs.
24/Cs.
384/Cs.
48/Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
02-992-428 60 mL 24/Cs.
02-992-955 30 mL 384/Cs.
02-992-449 30 mL 48/Cs.
02-992-436 120 mL 24/Cs.
02-992-787 180 mL 24/Cs.
02-992-706 240 mL 24/Cs.
02-992-415 480 mL 12/Cs.
02-992-406 960 mL 12/Cs.
02-993-294 60 mL 144/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02-992-428 Supplier Qorpak™ Supplier No. GLC01611
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Designed without shoulders for maximum storage capacity

  • Provides content visibility
  • Closure (Green thermoset (phenolic)) provides the chemical compatibility and most temperature tolerant
  • Foam backed PTFE liners provides resilience for tight seal with low moisture transmission rate

Analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.

Specifications

Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Color Clear
Diameter (English) 2.17 in.
Material Glass
Quantity 24/Cs.
Sterility Non-sterile
Closure Color Green
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Height (Metric) 49 mm
Height (English) 1.93 in.
Closure Liner F217, PTFE
Closure Material Thermoset
Closure Size 53-400 mm
Diameter (Metric) 55 mm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Capacity (English) 2 oz.
Shape Straight-sided
Product Type Wide Mouth Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.