Learn More
Qorpak™ Clear Straight Sided Round Bottles with Green Thermoset F217 and PTFE Caps
Description
- Provides content visibility
- Closure (Green thermoset (phenolic)) provides the chemical compatibility and most temperature tolerant
- Foam backed PTFE liners provides resilience for tight seal with low moisture transmission rate
Analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Specifications
Specifications
| Capacity | 1 oz., 30mL |
| Capacity (Metric) | 30 mL |
| Color | Clear |
| Diameter (English) | 1.69 in. |
| Material | Glass |
| Quantity | 48/Cs. |
| Sterility | Non-sterile |
| Closure Color | Green |
| Closure Included | Yes |
| Graduated | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.